missing translation for 'onlineSavingsMsg'
Learn More

Selenium ingot/button,≈38mm (1.5 in.) dia., Thermo Scientific Chemicals

Catalog Number 15422157
Click to view available options
Quantity:
1 Pc.
This item is not returnable. View return policy
This item is not returnable. View return policy

 

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS 7782-49-2
Physical Form Pieces
Quantity 1 Pc.
Molecular Formula Se
MDL Number MFCD00134090 MFCD00011224
Synonym selenium, elemental, selen, hydride, atom, elemental, selenio, vandex, alloy, and compounds, gray
InChI Key BUGBHKTXTAQXES-UHFFFAOYSA-N
SMILES [Se]
IUPAC Name selenium
Molecular Weight (g/mol) 78.97
PubChem CID 6326970
ChEBI CHEBI:27568
Odor Odorless
Chemical Name or Material Selenium ingot/button, ≈38mm (1.5 in.) dia.
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'