missing translation for 'onlineSavingsMsg'
Learn More

Selenite Lactose Broth

Reduce preparation time when enriching for Salmonella species from fecal and food samples with ready-to-use prepared media tubes.

Brand:  Thermo Scientific Oxoid EB0213E

Code : 64

Additional Details : Weight : 1.00000kg

 View more versions of this product

Product Code. 12753586

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


Specifications

Specifications

Light ivory, transparent
100mL
Universal Tube
100 universals in a carton box
2-10°C
Selenite F Broth
10 ± 0.5mL
Scotland
100/Pk.
Videos
Documentation

Technical Documents

For In Vitro Diagnostic Use.