missing translation for 'onlineSavingsMsg'
Learn More

SecinH3, Tocris Bioscience™

Sec7-specific GEF inhibitor (cytohesins)

Brand:  Tocris (Bio-Techne) 284910

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 10mg

 View more versions of this product

Product Code. 13461934

  • £211.20 / 10mg


Estimated Shipment
28-02-2019
Add to basket

Description

Description

Sec7-specific GEF inhibitor that displays selectivity for small GEFs of the cytohesin family (IC50 values are 2.4, 5.4, 5.4, 5.6, 5.6, 65 and > 100 μM for hCyh2, hCyh1, mCyh3, hCyh3, drosophila steppke, yGea2-S7 and hEFA6-S7 respectively). Inhibits insulin signaling via inhibition of insulin receptor substrate protein (IRS) phosphorylation.
Specifications

Chemical Identifiers

853625-60-2
460.508
secinh3, cytohesins inhibitor secinh3, secinh3 and cytohesins inhibitor secinh3, n-4-5-1,3-benzodioxol-5-yl-3-methoxy-1h-1,2,4-triazol-1-yl phenyl-2-phenylthio acetamide, n-4-5-1,3-benzodioxol-5-yl-3-methoxy-1h-1,2,4-triazol-1-yl phenyl-2-phenylthio acetamide, secin h3, n-4-5-benzo d 1,3 dioxol-5-yl-3-methoxy-1h-1,2,4-triazol-1-yl phenyl-2-phenylthio acetamide, n-4-5-1,3-benzodioxol-5-yl-3-methoxy-1,2,4-triazol-1-yl phenyl-2-phenylsulfanylacetamide, n-4-5-2h-1,3-benzodioxol-5-yl-3-methoxy-1h-1,2,4-triazol-1-yl phenyl-2-phenylsulfanyl acetamide
N-[4-[5-(1,3-benzodioxol-5-yl)-3-methoxy-1,2,4-triazol-1-yl]phenyl]-2-phenylsulfanylacetamide
C24H20N4O4S
QPGYAMIHXLCFTJ-UHFFFAOYSA-N
1029232
COC1=NN(C(=N1)C2=CC3=C(C=C2)OCO3)C4=CC=C(C=C4)NC(=O)CSC5=CC=CC=C5

Specifications

SecinH3
853625-60-2
secinh3, cytohesins inhibitor secinh3, secinh3 and cytohesins inhibitor secinh3, n-4-5-1,3-benzodioxol-5-yl-3-methoxy-1h-1,2,4-triazol-1-yl phenyl-2-phenylthio acetamide, n-4-5-1,3-benzodioxol-5-yl-3-methoxy-1h-1,2,4-triazol-1-yl phenyl-2-phenylthio acetamide, secin h3, n-4-5-benzo d 1,3 dioxol-5-yl-3-methoxy-1h-1,2,4-triazol-1-yl phenyl-2-phenylthio acetamide, n-4-5-1,3-benzodioxol-5-yl-3-methoxy-1,2,4-triazol-1-yl phenyl-2-phenylsulfanylacetamide, n-4-5-2h-1,3-benzodioxol-5-yl-3-methoxy-1h-1,2,4-triazol-1-yl phenyl-2-phenylsulfanyl acetamide
COC1=NN(C(=N1)C2=CC3=C(C=C2)OCO3)C4=CC=C(C=C4)NC(=O)CSC5=CC=CC=C5
460.508
460.5
10mg
C24H20N4O4S
QPGYAMIHXLCFTJ-UHFFFAOYSA-N
N-[4-[5-(1,3-benzodioxol-5-yl)-3-methoxy-1,2,4-triazol-1-yl]phenyl]-2-phenylsulfanylacetamide
1029232
>99%
Videos