missing translation for 'onlineSavingsMsg'
Learn More

sec-Butylbenzene, 99+%, ACROS Organics™

Brand:  Acros Organics 107862500

Code : 29

Additional Details : CAS Number : 135-98-8 Weight : 0.26500kg Transport : UN number : 2709 Chem class : 3 Pack group : III


Quantity 250mL
Packaging Glass bottle

 View more versions of this product

Product Code. 10579820

  • £46.70 / 250mL


EU Stock 2
Add to basket


Specifications

Chemical Identifiers

sec-Butylbenzene
135-98-8
99+%
C6H5CH(CH3)C2H5
MFCD00009329
99+%
≥99.0%
C10H14
134.22
2-Phenylbutane

Specifications

sec-Butylbenzene
135-98-8
Authentic
≥99.0%
Glass bottle
1.4890 to 1.4910 (20°C, 589nm)
134.22
05, 414
2-Phenylbutane
Clear
99+%
0.1% max. (GC)
Liquid
99+%
C10H14
C6H5CH(CH3)C2H5
MFCD00009329
15, 1552
0.05% max. (K.F.)
250mL
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • sec-Butylbenzene

Signal Word
  • Warning

Hazard Category
  • Flammable liquid Category 3

Hazard Statement
  • H226-Flammable liquid and vapour.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.

SDS