missing translation for 'onlineSavingsMsg'
Learn More

sec-Butanol, 99+%, for analysis, Thermo Scientific Chemicals

Catalog Number p-3759001
Click to view available options
Quantity:
1 L
2.5 L
Packaging:
Glass Bottle
78-92-2
C4H10O
74.12
MFCD00004569
BTANRVKWQNVYAZ-UHFFFAOYNA-N
2-butanol, sec-butanol, sec-butyl alcohol, 2-hydroxybutane, 2-butyl alcohol, s-butyl alcohol, butylene hydrate, s-butanol, 1-methyl-1-propanol, 1-methylpropyl alcohol
6568
CHEBI:35687
butan-2-ol
CCC(C)O
78-92-2
C4H10O
74.12
MFCD00004569
BTANRVKWQNVYAZ-UHFFFAOYNA-N
2-butanol, sec-butanol, sec-butyl alcohol, 2-hydroxybutane, 2-butyl alcohol, s-butyl alcohol, butylene hydrate, s-butanol, 1-methyl-1-propanol, 1-methylpropyl alcohol
6568
CHEBI:35687
butan-2-ol
CCC(C)O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point -115.0°C
Density 0.8100g/mL
Boiling Point 99.0°C
Quantity 1 L
Flash Point 24°C
Packaging Glass Bottle
Physical Form Liquid
Assay Percent Range 99+%
Specific Gravity 0.81
Linear Formula C2H5CH(OH)CH3
Viscosity 4.2 mPa.s (20°C)
Beilstein 01, 371
Water 0.1% max.
Merck Index 15, 1543
Solubility Information Solubility in water: 125g/L (20°C). Other solubilities: soluble in alcohol, ester, ether and benzene
Formula Weight 74.12
Percent Purity ≥99%
Grade Analytical
Chemical Name or Material sec-Butanol, For analysis
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'