missing translation for 'onlineSavingsMsg'
Learn More

Sebacoyl chloride, 95%, Thermo Scientific Chemicals

Catalog Number p-7021758
Click to view available options
Quantity:
25 g
100 g
500 g
111-19-3
C10H16Cl2O2
239.136
MFCD00000770
WMPOZLHMGVKUEJ-UHFFFAOYSA-N
sebacoyl chloride, sebacyl chloride, sebacoyl dichloride, sebacic acid dichloride, decanedioyl chloride, sebacoylchloride, octane-1,8-dicarbonyl chloride, sebacoyl chloride, cp, acmc-1byrk, decanedioic acid chloride
66072
decanedioyl dichloride
C(CCCCC(=O)Cl)CCCC(=O)Cl
111-19-3
C10H16Cl2O2
239.136
MFCD00000770
WMPOZLHMGVKUEJ-UHFFFAOYSA-N
sebacoyl chloride, sebacyl chloride, sebacoyl dichloride, sebacic acid dichloride, decanedioyl chloride, sebacoylchloride, octane-1,8-dicarbonyl chloride, sebacoyl chloride, cp, acmc-1byrk, decanedioic acid chloride
66072
decanedioyl dichloride
C(CCCCC(=O)Cl)CCCC(=O)Cl

Sebacoyl chloride is used in the synthesis of nylon-6,10 polymer by reacting with hexamethylenediamine. It is also used to prepare decanedioic acid bis-(3-dimethylamino-propyl) ester in the presence of triethylamine as catalyst. It is used as an intermediate in chemical synthesis. It is involved in the production of polyamide microcapsules by polycondensation of p-phenylenediamine with sebacoyl chloride using diethylenetriamine (DETA) cross-linker and oleic acid as a plasticizer.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sebacoyl chloride is used in the synthesis of nylon-6,10 polymer by reacting with hexamethylenediamine. It is also used to prepare decanedioic acid bis-(3-dimethylamino-propyl) ester in the presence of triethylamine as catalyst. It is used as an intermediate in chemical synthesis. It is involved in the production of polyamide microcapsules by polycondensation of p-phenylenediamine with sebacoyl chloride using diethylenetriamine (DETA) cross-linker and oleic acid as a plasticizer.

Solubility
Miscible with hydrocarbons and ethers.

Notes
Store in a cool place. Moisture sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with water, acids, bases, amines, alcohols and oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point -5°C
Density 1.121
Boiling Point 161°C (14 mmHg)
Flash Point 165°C (329°F)
Odor Acrid
Refractive Index 1.468
Quantity 25 g
UN Number UN2922
Beilstein 1365665
Sensitivity Moisture sensitive
Solubility Information Miscible with hydrocarbons and ethers.
Formula Weight 239.14
Percent Purity 97%
Chemical Name or Material Sebacoyl chloride
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'