missing translation for 'onlineSavingsMsg'
Learn More

Sebacic Acid, 98%, ACROS Organics™

Brand:  Acros Organics 132630025

Code : 29

Additional Details : CAS Number : 111-20-6 Weight : 2.55000kg

Quantity 2.5kg
Packaging Plastic bottle

 View more versions of this product

Product Code. 10667292

  • £97.80 / 2.50kg


EU Stock 3
Add to basket

Specifications

Chemical Identifiers

Sebacic acid
98%
HO2C(CH2)8CO2H
MFCD00004440
98%
C10H18O4
202.25
Decanedioic acid, 1, 8-Octanedicarboxylic acid

Specifications

Sebacic acid
0.06% max.
98%
C10H18O4
HO2C(CH2)8CO2H
MFCD00004440
15, 8552
Decanedioic acid, 1, 8-Octanedicarboxylic acid
0.3% max.
White to off-white
2.5kg
111-20-6
Powder or Granules
98%
Authentic
202.25
02, IV, 2078
Plastic bottle
Solubility in water: 1g/L (20°C). Other solubilities: 0.1% in ether (17°C), freely soluble in alcohol, esters and ketones, sparingly soluble in hydrocarbons, insoluble in ethyl ether and carbon tetrachloride
294.4°C (100.0 mmHg)
131.0° to 134.0°C
220°C
Videos
SDS