missing translation for 'onlineSavingsMsg'
Learn More

Sebacic Acid, 98%, Thermo Scientific Chemicals

Catalog Number p-3737360
Click to view available options
Quantity:
100 g
1 kg
2.5 kg
10 kg
Packaging:
Plastic bottle
Plastic drum
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
111-20-6
C10H18O4
202.25
MFCD00004440
CXMXRPHRNRROMY-UHFFFAOYSA-N
sebacic acid, 1,8-octanedicarboxylic acid, 1,10-decanedioic acid, sebacic acids, decanedicarboxylic acid, n-decanedioic acid, sebacinsaure, acide sebacique, sebacate, sebacinsaeure
5192
CHEBI:41865
decanedioic acid
C(CCCCC(=O)O)CCCC(=O)O
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
111-20-6
C10H18O4
202.25
MFCD00004440
CXMXRPHRNRROMY-UHFFFAOYSA-N
sebacic acid, 1,8-octanedicarboxylic acid, 1,10-decanedioic acid, sebacic acids, decanedicarboxylic acid, n-decanedioic acid, sebacinsaure, acide sebacique, sebacate, sebacinsaeure
5192
CHEBI:41865
decanedioic acid
C(CCCCC(=O)O)CCCC(=O)O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 131.0°C to 134.0°C
Color White
Boiling Point 294.4°C (100.0 mmHg)
Flash Point 220°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Plastic bottle
Linear Formula HO2C(CH2)8CO2H
Quantity 100 g
Beilstein 02, IV, 2078
Merck Index 15, 8552
Solubility Information Solubility in water: 1g/L (20°C). Other solubilities: 0.1% in ether (17°C), freely soluble in alcohol, esters and ketones, sparingly soluble in hydrocarbons, insoluble in ethyl ether and carbon tetrachloride
Formula Weight 202.25
Percent Purity 98%
Physical Form Powder or Granules
Chemical Name or Material Sebacic acid
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'