missing translation for 'onlineSavingsMsg'
Learn More

SDZ SER 082 fumarate, Tocris Bioscience™

Selective 5-HT2B/2C antagonist

Brand:  Tocris (Bio-Techne) 125550

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13428993

  • £593.88 / 50mg

Estimated Shipment date
from Supplier 03-04-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Selective 5-HT2B/2C receptor antagonist with low affinity for 5-HT1A receptors. Inhibits [3H]-mesulergine binding to 5-HT2C receptors with a pKD of 7.8. Inhibits 5-HT2B mediated responses in the rat fundus (pKB = 7.34).
Specifications

Chemical Identifiers

1417343-80-6
344.411
sdz ser 082 fumarate, sdz ser 082 fumarate , 141474-54-6 sdz ser 082 ., +-cis-4,5,7a,8,9,10,11,11a-octahydro-7h-10-methylindolo 1,7-bc 2,6-naphthyridine fumarate
CN1CCC2CN3CCC4=C3C(=CC=C4)C2C1.C(=CC(=O)O)C(=O)O
C19H24N2O4
DXUZZRDHJMOLTN-MRYVXRNOSA-N
45073480

Specifications

50mg
1417343-80-6
sdz ser 082 fumarate, sdz ser 082 fumarate , 141474-54-6 sdz ser 082 ., +-cis-4,5,7a,8,9,10,11,11a-octahydro-7h-10-methylindolo 1,7-bc 2,6-naphthyridine fumarate
CN1CCC2CN3CCC4=C3C(=CC=C4)C2C1.C(=CC(=O)O)C(=O)O
45073480
>98%
SDZ SER 082 fumarate
C19H24N2O4
DXUZZRDHJMOLTN-MRYVXRNOSA-N
344.411
344.41
Videos
Documents