missing translation for 'onlineSavingsMsg'
Learn More

SDZ 220-040, Tocris Bioscience™

Potent, competitive NMDA antagonist

Brand:  Tocris (Bio-Techne) 125110

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 10mg

 View more versions of this product

Product Code. 13438983

  • £163.17 / 10mg

Estimated Shipment date
from Supplier 20-11-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Potent competitive antagonist at the NMDA receptor (pKi = 8.5). Selective over a range of other receptor sites.
Specifications

Chemical Identifiers

174575-40-7
420.179
s-2-amino-3-2',4'-dichloro-4-hydroxy-5-phosphonomethyl-1,1'-biphenyl-3-yl propanoic acid, tocris-1251, 2s-2-amino-3-5-2,4-dichlorophenyl-2-hydroxy-3-phosphonomethyl phenyl propanoic acid, 2s-2-amino-3-2',4'-dichloro-4-hydroxy-5-phosphonomethyl-1,1'-biphenyl-3-yl propanoic acid, s-?-amino-2',4'-dichloro-4-hydroxy-5-phosphonomethyl-1,1'-biphenyl-3-propanoic acid, 1,1'-biphenyl-3-propanoicacid,a-amino-2',4'-dichloro-4-hydroxy-5-phosphonomethyl-, as
(2S)-2-amino-3-[5-(2,4-dichlorophenyl)-2-hydroxy-3-(phosphonomethyl)phenyl]propanoic acid
C16H16Cl2NO6P
NYZFUZCCDOSQBG-AWEZNQCLSA-N
6604839
C1=CC(=C(C=C1Cl)Cl)C2=CC(=C(C(=C2)CP(=O)(O)O)O)CC(C(=O)O)N

Specifications

10mg
174575-40-7
s-2-amino-3-2',4'-dichloro-4-hydroxy-5-phosphonomethyl-1,1'-biphenyl-3-yl propanoic acid, tocris-1251, 2s-2-amino-3-5-2,4-dichlorophenyl-2-hydroxy-3-phosphonomethyl phenyl propanoic acid, 2s-2-amino-3-2',4'-dichloro-4-hydroxy-5-phosphonomethyl-1,1'-biphenyl-3-yl propanoic acid, s-?-amino-2',4'-dichloro-4-hydroxy-5-phosphonomethyl-1,1'-biphenyl-3-propanoic acid, 1,1'-biphenyl-3-propanoicacid,a-amino-2',4'-dichloro-4-hydroxy-5-phosphonomethyl-, as
C1=CC(=C(C=C1Cl)Cl)C2=CC(=C(C(=C2)CP(=O)(O)O)O)CC(C(=O)O)N
420.179
420.19
SDZ 220-040
C16H16Cl2NO6P
NYZFUZCCDOSQBG-AWEZNQCLSA-N
(2S)-2-amino-3-[5-(2,4-dichlorophenyl)-2-hydroxy-3-(phosphonomethyl)phenyl]propanoic acid
6604839
>98%
Videos
Documents