missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SDM25N hydrochloride, Tocris Bioscience™
Potent, selective non-peptide δ antagonist; also inhibits dengue virus replication
Brand: Tocris (Bio-Techne) 141050
Code : Z1
Additional Details : Weight : 0.01000kg
Quantity | 50mg |
---|
Description
Potent and highly selective non-peptide δ receptor antagonist (Ki values are 4.7, 3800 and 7900 nM for δ, κ and μ receptors respectively). More selective than naltrindole (Cat. No. 0740). Also inhibits Dengue virus replication.Chemical Identifiers
342884-71-3 | |
450.963 | |
sdm25n hydrochloride, 4bs,8r,8as,14br-5,6,7,8,14,14b-hexahydro-7-2-methyl-2-propenyl-4,8-methanobenzofuro 2,3-a pyrido 4,3-b carbazole-1,8a 9h-diol hydrochloride | |
CC(=C)CN1CCC23C4C5=C(CC2(C1CC6=C3C(=C(C=C6)O)O4)O)C7=CC=CC=C7N5.Cl |
C26H27ClN2O3 | |
QLLCUVACGPLGAX-IDRHMUJXSA-N | |
122360814 |
Specifications
SDM25N hydrochloride | |
342884-71-3 | |
sdm25n hydrochloride, 4bs,8r,8as,14br-5,6,7,8,14,14b-hexahydro-7-2-methyl-2-propenyl-4,8-methanobenzofuro 2,3-a pyrido 4,3-b carbazole-1,8a 9h-diol hydrochloride | |
CC(=C)CN1CCC23C4C5=C(CC2(C1CC6=C3C(=C(C=C6)O)O4)O)C7=CC=CC=C7N5.Cl | |
122360814 | |
>99% |
50mg | |
C26H27ClN2O3 | |
QLLCUVACGPLGAX-IDRHMUJXSA-N | |
450.963 | |
450.96 |