missing translation for 'onlineSavingsMsg'
Learn More

SCYL3 Monoclonal Antibody (OTI4F10), Invitrogen™

Mouse Monoclonal Antibody

Brand:  Thermo Scientific MA524934

Additional Details : Weight : 0.01000kg

Product Code. 15904544

  • £432.00 / 100µL
Estimated Shipment: 23-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

SCYL3 Monoclonal antibody specifically detects SCYL3 in Human, Non-human Primate samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.
Specifications

Specifications

SCYL3
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Q8IZE3
SCYL3
Full-length protein expressed in 293T cell transfected with human SCYL3 expression vector
100 μL
Primary
Human, Non-human Primate
Antibody
IgG1
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
OTI4F10
Unconjugated
SCYL3
1200016D23Rik; 6030457O16; AW214499; ezrin-binding partner PACE-1; ezrin-binding partner PACE-1 (PACE-1); ezrin-binding protein PACE-1; Pace1; PACE-1; protein-associating with the carboxyl-terminal domain of ezrin; RP1-97P20.2; SCY1 like pseudokinase 3; SCY1-like 3; SCY1-like 3 (S. cerevisiae); SCY1-like protein 3; SCY1-like, kinase-like 3; Scyl3
Mouse
Affinity Chromatography
RUO
57147
-20° C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers