Learn More
Tocris Bioscience™ Scriptaid
Histone deacetylase inhibitor
Brand:  Tocris Bioscience™ 242150
Code : Z1
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 13431184
  • £552.04 / 50mg
Estimated Shipment
14-09-2020
Description
Description
Novel histone deacetylase inhibitor. Produces > 100-fold increase in histone acetylation and displays relatively low toxicity.
Specifications
Specifications
Scriptaid
>99
50mg
Scriptaid
326.35