Learn More
Thermo Scientific™ 8mm Autosampler Vial Septa
Thermo Scientific™ Septa for 8-425 Screw Thread Caps are constructed of PTFE, PTFE/rubber, or PTFE/silicone to suit a variety of HPLC, GC and IC applications.
Brand: Thermo Scientific™ 60180-718
Code : 16
Additional Details : Weight : 1.00000kg
Description
Thermo Scientific™ Septa for 8-425 Screw Thread Caps are constructed of PTFE, PTFE/rubber, or PTFE/silicone to suit a variety of HPLC, GC and IC applications.
Specifications
White/Red | |
PTFE, Silicone | |
Thermo Scientific Spectra System AS3500/AS3000/AS1000/Surveyor/Accela LC, Agilent 1090, PE Autosystem |
Red PTFE/white silicone seal; Thermo Scientific, Agilent, Claris, Shimadzu, Varian, Waters, Hitachi | |
Red PTFE/white silicone seal |