Learn More
JG Finneran Associates™ Screw Caps
Use these preassembled caps for a variety of applications. JG Finneran™ JG Finneran™ Screw Caps provide a positive physical lock to assure that preassembled components arrive intact.
Brand: JG Finneran Associates™ 806550TH-08
Code : 17
Additional Details : Weight : 0.06204kg
Description
- The patented Top Hat™ septa and closure provide a positive physical lock to assure that preassembled components arrive intact
- Preassembled R.A.M.™ Smooth Screw Thread Closure and Septa
- Designed to work in Agilent or other robotic arm autosamplers
- Available in a variety of colors
Specifications
Black | |
PTFE/Silicone | |
Screw Cap |
PTFE, Silicone | |
100 Pk. | |
8-425 |