missing translation for 'onlineSavingsMsg'
Learn More

Kimberly-Clark™ SCOTT™ XTRA Interfolded Hand Towels

Provides efficient and hygienic hand drying. Kimberly-Clark™ SCOTT™ XTRA Interfolded Hand Towels are interfolded for one-at-a-time easy dispensing.

Brand:  Kimberly-Clark™ 6679

Code : 02

Additional Details : Weight : 1.00000kg

Product Code. 12754326

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


Description

Description

  • White in color
  • Soft yet high absorbent, even when wet
  • Made using unique AIRFLEX™ technology
  • Ideal for high-traffic areas where best-in-class cost efficiency is critical
Specifications

Specifications

White
Folded
21.5 cm
Paper
21 x 21.5 cm
3750/Pk.
21 cm
Hand Towel
Videos