missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SCN1A Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA567968
Additional Details : Weight : 0.01000kg
Description
Predicted to react with Mouse and Rat samples.
SCN1A Polyclonal antibody specifically detects SCN1A in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
SCN1A | |
Polyclonal | |
Unconjugated | |
Scn1a | |
3-Feb; B230332M13; BFIC3; BFIS3; BFNIS; brain III voltage-gated sodium channel; Brain Type I; cardiac tetrodotoxin-insensitive voltage-dependent sodium channel alpha subunit; CDCD2; CERIII; CIAT; CIN5; CMD1E; CMPD2; CMS16; CTC-264K15.6; EIEE11; EIEE13; EIEE6; ETHA; FEB3; FEB3A; FEB3B; FEPS2; FEPS3; FHM3; GEFS+2; GEFSP2; GEFSP7; HB1; HB2; HBA; HBBD; HBSC I; HBSC II; HBSCI; HBSCII; HH1; hNa6/Scn8a voltage-gated sodium channel; hNaN; hNE-Na; HOKPP2; hPN3; HSAN2D; HSAN7; HYKPP; HYPP; ICCD; IVF; KIAA1356; LQT3; MED; Na(v)1.2; Na(V)1.4; NAC1; NAC1A; NAC2; NAC3; NaCh6; NaN; Nav1.1; Nav1.2; Nav1.3; Nav1.4; Nav1.5; Nav1.6; Nav1.7; Nav1.8; NAV1.9; NENA; NE-NA; Neuroendocrine sodium channel; peripheral nerve sodium channel 3; Peripheral nerve sodium channel 5; peripheral sodium channel 1; PFHB1; PN1; PN3; PN4; PN5; RBI; SCN1; SCN10A; SCN11A; SCN12A; Scn1a; SCN2A; SCN2A1; SCN2A2; SCN3A; SCN4A; SCN5A; SCN8A; SCN9A; Sensory neuron sodium channel 2; SFNP; skeletal muscle sodium channel alpha subunit; | |
Rabbit | |
affinity chromatography | |
RUO | |
20265, 6323, 81574 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
A2APX8, P04774, P35498 | |
Scn1a | |
Synthesized peptide derived from human SCN1A. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |