Learn More
Kartell™ Schilling Burette
Automatic plastic Schilling burette (according to din 12.700 standards, Class B). With push button for drop-by-drop delivery. PE reservoir with stable PE-LD base.
Brand:  Kartell™ 0198100
Code : NEW
Additional Details : Weight : 0.50000kg
 View more versions of this product
Product Code. 10748104
  • £143.00 / Each
Estimated Shipment: 18-07-2024
to see stock.
Add to basket
Description
Description
Automatic plastic Schilling burette (according to din 12.700 standards, Class B). With push button for drop-by-drop delivery. PE reservoir with stable PE-LD base.
  • Automatic plastic Schilling burettes (according to din 12.700 standards, Class B)
  • With push button for drop-by- drop delivery
  • PE reservoir with stable PE-LD base
  • Specifications
    Specifications
    50 mL
    Schilling Buret
    DIN 12700
    PMP, Polyethylene
    Class B