missing translation for 'onlineSavingsMsg'
Learn More

SCH 50911, Tocris Bioscience™

Selective and competitive GABAB antagonist; orally bioavailable

Brand:  Tocris (Bio-Techne) 098450

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13438233

  • £782.92 / 50mg

Estimated Shipment
11-09-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

A selective, competitive and orally active GABAB antagonist. Displays an IC50 of 1.1 μM at GABAB, approximately 60 times that of CGP 35348 (Cat. No. 1245) and no binding affinity for GABAA at concentrations up to 100 μM.
Specifications

Chemical Identifiers

733717-87-8
173.212
sch-50911 free base, unii-13ob0keu61, 2s-5,5-dimethylmorpholin-2-yl acetic acid, dsstox_cid_25670, dsstox_rid_81045, 2s-+-5,5-dimethyl-2-morpholineacetic acid, dsstox_gsid_45670, 2-2s-5,5-dimethylmorpholin-2-yl acetic acid, spectrum_001764, sch 50911 free base
2-[(2S)-5,5-dimethylmorpholin-2-yl]acetic acid
C8H15NO3
SEYCKMQSPUVYEF-LURJTMIESA-N
5311429
CC1(COC(CN1)CC(=O)O)C

Specifications

SCH 50911
733717-87-8
sch-50911 free base, unii-13ob0keu61, 2s-5,5-dimethylmorpholin-2-yl acetic acid, dsstox_cid_25670, dsstox_rid_81045, 2s-+-5,5-dimethyl-2-morpholineacetic acid, dsstox_gsid_45670, 2-2s-5,5-dimethylmorpholin-2-yl acetic acid, spectrum_001764, sch 50911 free base
CC1(COC(CN1)CC(=O)O)C
173.212
173.21
50mg
C8H15NO3
SEYCKMQSPUVYEF-LURJTMIESA-N
2-[(2S)-5,5-dimethylmorpholin-2-yl]acetic acid
5311429
>98%
Videos
Documents