missing translation for 'onlineSavingsMsg'
Learn More

SCH 28080, Tocris Bioscience™

H+,K+-ATPase inhibitor

Brand:  Tocris (Bio-Techne) 169050

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13459923

  • £517.44 / 50mg

Estimated Shipment date
from Supplier 30-03-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Potent inhibitor of H+,K+-ATPase (IC50 = 20 nM); binds to the K+ recognition site and is competitive with respect to K+. Inhibits gastric acid secretion in vitro and in vivo.
Specifications

Chemical Identifiers

76081-98-6
277.327
schering compound 28080, 2-methyl-8-phenylmethoxy imidazo 1,2-a pyridine-3-acetonitrile, chembl47529, 2-methyl-8-phenylmethoxy imidazo-1-2-a pyrine-3-acetonitrile, 2-8-benzyloxy-2-methylimidazo 1,2-a pyridin-3-yl acetonitrile, imidazo 1,2-a pyridine-3-acetonitrile, 2-methyl-8-phenylmethoxy, tocris-1690, 8-benzyloxy-2-methylimidazo 1,2-a pyridin-3-yl acetonitrile, lopac-s-4443
2-(2-methyl-8-phenylmethoxyimidazo[1,2-a]pyridin-3-yl)acetonitrile
C17H15N3O
PYKJFEPAUKAXNN-UHFFFAOYSA-N
108137
CC1=C(N2C=CC=C(C2=N1)OCC3=CC=CC=C3)CC#N

Specifications

50mg
76081-98-6
schering compound 28080, 2-methyl-8-phenylmethoxy imidazo 1,2-a pyridine-3-acetonitrile, chembl47529, 2-methyl-8-phenylmethoxy imidazo-1-2-a pyrine-3-acetonitrile, 2-8-benzyloxy-2-methylimidazo 1,2-a pyridin-3-yl acetonitrile, imidazo 1,2-a pyridine-3-acetonitrile, 2-methyl-8-phenylmethoxy, tocris-1690, 8-benzyloxy-2-methylimidazo 1,2-a pyridin-3-yl acetonitrile, lopac-s-4443
CC1=C(N2C=CC=C(C2=N1)OCC3=CC=CC=C3)CC#N
277.327
277.33
SCH 28080
C17H15N3O
PYKJFEPAUKAXNN-UHFFFAOYSA-N
2-(2-methyl-8-phenylmethoxyimidazo[1,2-a]pyridin-3-yl)acetonitrile
108137
>99%
Videos
Documents