Sino Biological™ SCGN Recombinant Human Protein, N-His Tag

Expressed from DNA sequence encoding human SCGN (Asp 2-Pro 276) fused to polyhistidinetag at N-terminus

Brand: Sino Biological™

Manufacturer Part Number: 12411H07E25


UNSPSC: 12352202

Code: NEW

Additional Details:
Additional Details: Weight: 0.01000kg

Disclaimers: For Research Use Only. Not for use in diagnostic procedures.

Product Code. 15235915

Quantity Price
1 £133.00 / 25µg
Estimated Shipment date
from Supplier 27-10-2016
Add to basket

Description and Specification


For Use With (Application) Analysis of protein stucture and used as immunogen, protein standard, or in cell biology research applications
Format Lyophilized
Gene ID (Entrez) 10590
Molecular Weight 33.4kd
Purity ≥90% by SDS-PAGE
Quantity 5 x 5μg
Source Human
Structural Form Full Length, Recombinant
Endotoxin Concentration <1 EU/μg
Common Name Secretagogin, Ef-Hand Calcium Binding Protein
Gene Symbol SCGN
Protein Tag His Tag
Validated Application Assay Standard
Protein Family Other Proteins
Expression System E.coli
Product Type SCGN Recombinant Human Protein

  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80