missing translation for 'onlineSavingsMsg'
Learn More

Sino Biological™ SCGN Recombinant Human Protein, N-His Tag

Expressed from DNA sequence encoding human SCGN (Asp 2-Pro 276) fused to polyhistidinetag at N-terminus

Brand:  Sino Biological™ 12411H07E50

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15265915

  • £213.41 / 50µg


Estimated Shipment
09-02-2018
Add to basket

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Description

Description

  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
Specifications

Specifications

Analysis of protein stucture and used as immunogen, protein standard, or in cell biology research applications
10590
≥90% by SDS-PAGE
Human
<1 EU/μg
SCGN
Other Proteins
SCGN Recombinant Human Protein
Lyophilized
33.4kd
50μg
Full Length, Recombinant
Secretagogin, Ef-Hand Calcium Binding Protein
Assay Standard
E.coli
His Tag
Videos