missing translation for 'onlineSavingsMsg'
Learn More

Sino Biological™ SCGB1A1 Recombinant Mouse Protein, His Tag

Expressed from DNA sequence encoding mouse SCGB1A1 (Q06318) (Met 1-Phe 96) fused to polyhistidine tag at the C-terminus

Brand:  Sino Biological™ 50291M08H50

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15373066

  • £220.70 / 50µg


Estimated Shipment
15-06-2018
Add to basket

Description

Description

  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
Specifications

Specifications

Analysis of protein stucture and used as immunogen, protein standard, or in cell biology research applications
22287
≥85% by SDS-PAGE
Mouse
<1 EU/μg
SCGB1A1
Other Proteins
SCGB1A1 Recombinant Mouse Protein
Lyophilized
9.8kd
50μg
Full Length, Recombinant
Secretoglobin, Family 1A, Member 1 (Uteroglobin)
Assay Standard
Mammalian
His Tag
Videos

For Research Use Only. Not for use in diagnostic procedures.