Learn More
SCD5 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA559963
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: NTQHIQKEGR ALNQEAACEM LREWHQGHIL KVTLPGLHIL ALLHTHCNHS EKCCLMLRAL SVSLEVF Highest antigen sequence identity to the following orthologs: Mouse - 24%, Rat - 23%.
Stearoyl-CoA desaturase is an integral membrane protein of the endoplasmic reticulum that catalyzes the formation of monounsaturated fatty acids from saturated fatty acids. SCD may be a key regulator of energy metabolism with a role in obesity and dislipidemia. Four SCD isoforms, Scd1 through Scd4, have been identified in mouse. In contrast, only 2 SCD isoforms, SCD1 and SCD5, have been identified in human. SCD1 shares about 85% amino acid identity with all 4 mouse SCD isoforms, as well as with rat Scd1 and Scd2. In contrast, SCD5 shares limited homology with the rodent SCDs and appears to be unique to primates.Specifications
SCD5 | |
Polyclonal | |
Unconjugated | |
SCD5 | |
ACOD4; acyl-CoA-desaturase 4; FADS4; HSCD5; SCD2; SCD4; SCD5; Stearoyl-CoA 9-desaturase; stearoyl-CoA desaturase 2; stearoyl-CoA desaturase 4; stearoyl-CoA desaturase 5 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
79966 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q86SK9 | |
SCD5 | |
Recombinant protein corresponding to Human SCD5 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only