missing translation for 'onlineSavingsMsg'
Learn More

Scandium(III) nitrate hydrate, REacton™, 99.9% (REO), Thermo Scientific Chemicals

Catalog Number p-7080290
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
107552-14-7
N3O9Sc
MFCD00011220
DFCYEXJMCFQPPA-UHFFFAOYSA-N
scandium nitrate, scandium iii nitrate, pa4cjgprtn, unii-pa4cjgprtn, scandium iii nitrate pentahydrate-sc reo, nitric acid, scandium 3+ salt 3:1, scandium 3+ trinitrate, acmc-1bol9, nitric acid, scandium salt
166818
scandium(3+);trinitrate
This item is not returnable. View return policy
107552-14-7
N3O9Sc
MFCD00011220
DFCYEXJMCFQPPA-UHFFFAOYSA-N
scandium nitrate, scandium iii nitrate, pa4cjgprtn, unii-pa4cjgprtn, scandium iii nitrate pentahydrate-sc reo, nitric acid, scandium 3+ salt 3:1, scandium 3+ trinitrate, acmc-1bol9, nitric acid, scandium salt
166818
scandium(3+);trinitrate

Scandium(III) nitrate hydrate is used as an oxidizing agent in organic synthesis. It is used in the optical coatings, electronic ceramics and laser industries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color White
Physical Form Crystalline
Quantity 1 g
UN Number UN1477
Merck Index 14,8392
Solubility Information Soluble in water and alcohol.
SMILES [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[Sc+3]
Molecular Weight (g/mol) 230.968
Formula Weight 230.97 (Anhydrous)
Percent Purity 99.9%
Grade Reacton™
Sensitivity Hygroscopic
Assay (REO)
Chemical Name or Material Scandium (III) nitrate hydrate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'