missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Scafftag™ Polypropylene Unitag Inserts

Inserts for Unitag™ System.

Brand:  Brady™ 806795

Code : 50

Additional Details : Weight : 0.00100kg

 View more versions of this product

Product Code. 12903025

  • £21.75 / 1 pack
Estimated Shipment: 14-05-2024
to see stock.
Add to basket

Description

Description

  • Inserts for Unitag™ System.
Specifications

Specifications

Unitag Insert
Polypropylene
Next Inspection Date Due
20 UniTag Inserts
-10°C to +90°C
White on Red
41 mm
Inspection tagging, Electrical labelling, Safety Identification
1 Ea.
0.5 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers