missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Scafftag™ Polypropylene Multitag Inserts

Inserts for Multitag™ System.

Brand:  Brady™ 806747

Code : 50

Additional Details : Weight : 0.00100kg

 View more versions of this product

Product Code. 12933005

  • £18.25 / 1 pack
Estimated Shipment: 15-05-2024
to see stock.
Add to basket

Description

Description

  • Inserts for Multitag™ System.
Specifications

Specifications

Multi-tag Insert
Polypropylene
Inspection tagging, Component and equipment labelling
164 mm
10 Pk.
0.5 mm
50 mm
Black on Yellow, White on Green
Equipment Inspection Record
6.46 in.
10 Multitag Inserts
-10°C to +90°C
1.97 in.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers