missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Scafftag Polypropylene Forklifttag Insert

Brand:  Brady™ 806699

Code : 50

Additional Details : Weight : 0.00100kg

 View more versions of this product

Product Code. 12982985

  • POA / 1 pack

This item cannot be ordered online because the price is subject to change. Please contact our Product Support team on 01509 555500 for assistance.


Specifications

Specifications

Black on Blue & Green
Inserts for use with Forklifttag™ System
10Pack
0.5mm
Forkliftag Insert
144mm
Daily Checklist
193mm
–10 to 90°C
Polypropylene
Forkliftag Insert
Videos