Learn More
Brady™ Scafftag™ Polypropylene Chemtag Inserts
Inserts for the Chemtag™ System. They contain all the pertinent information specific to the risks that hazardous substances pose, such as: substance type, details of hazard and precautions, supplier's name and emergency phone number, etc.
Brand: Brady™ 806683
Code : 50
Additional Details : Weight : 0.00100kg
Description
- Contain all the pertinent information specific to the risks that hazardous substances pose, such as:
- Substance type
- Details of hazard and precautions
- Supplier's name and emergency phone number
Specifications
Chemtag Insert | |
Polypropylene | |
Inspection tagging, Hazard communication Identification | |
193 mm | |
1 Ea. | |
-10°C to +90°C | |
5.67 in. |
Multicolor | |
Chemical Details | |
7.6 in. | |
50 ChemTag Inserts | |
14°F to +194°F | |
0.5 mm | |
144 mm |