missing translation for 'onlineSavingsMsg'
Learn More

SC 51322, Tocris Bioscience™

Potent EP1 receptor antagonist

Brand:  Tocris (Bio-Techne) 279110

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 10mg

 View more versions of this product

Product Code. 13451844

  • £176.40 / 10mg

Estimated Shipment date
from Supplier 31-03-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Potent EP1 prostanoid receptor antagonist (Ki = 13.8 nM). Displays analgesic properties in vivo.
Specifications

Chemical Identifiers

146032-79-3
457.929
8-chloro-2-3-2-furanylmethyl thio-1-oxopropyl-dibenz z b,f 1,4 oxazepine-10 11h-carboxylic acid hydrazide, d0v9qt, sc hplc, 3-chloro-n'-3-furan-2-ylmethylsulfanyl propanoyl-6h-benzo b 1,5 benzoxazepine-5-carbohydrazide, 6-chloro-n'-3-furan-2-ylmethyl sulfanyl propanoyl-2-oxa-9-azatricyclo 9.4.0.0 3 ,? pentadeca-1 15 ,3,5,7,11,13-hexaene-9-carbohydrazide, 8-chloro-dibenz b,f 1,4 oxazepine-10 11h-carboxylic acid 2-3-2-furanylmethyl thio-1-oxopropyl hydrazide, 8-chlorodibenz b,f 1,4 oxazepine-10 11h-carboxylic acid, 2-3-2-furanylmethyl thio-1-oxopropyl hydrazide, 8-chlorodibenz b,f 1,41oxazepine-10 11h-carboxylic acid, 2-3-2-furanylmethyl thio-1-oxopropyl hydrazide
3-chloro-N'-[3-(furan-2-ylmethylsulfanyl)propanoyl]-6H-benzo[b][1,4]benzoxazepine-5-carbohydrazide
C22H20ClN3O4S
CQBVTZDISUKDSX-UHFFFAOYSA-N
9933831
C1C2=CC=CC=C2OC3=C(N1C(=O)NNC(=O)CCSCC4=CC=CO4)C=C(C=C3)Cl

Specifications

SC 51322
146032-79-3
8-chloro-2-3-2-furanylmethyl thio-1-oxopropyl-dibenz z b,f 1,4 oxazepine-10 11h-carboxylic acid hydrazide, d0v9qt, sc hplc, 3-chloro-n'-3-furan-2-ylmethylsulfanyl propanoyl-6h-benzo b 1,5 benzoxazepine-5-carbohydrazide, 6-chloro-n'-3-furan-2-ylmethyl sulfanyl propanoyl-2-oxa-9-azatricyclo 9.4.0.0 3 ,? pentadeca-1 15 ,3,5,7,11,13-hexaene-9-carbohydrazide, 8-chloro-dibenz b,f 1,4 oxazepine-10 11h-carboxylic acid 2-3-2-furanylmethyl thio-1-oxopropyl hydrazide, 8-chlorodibenz b,f 1,4 oxazepine-10 11h-carboxylic acid, 2-3-2-furanylmethyl thio-1-oxopropyl hydrazide, 8-chlorodibenz b,f 1,41oxazepine-10 11h-carboxylic acid, 2-3-2-furanylmethyl thio-1-oxopropyl hydrazide
C1C2=CC=CC=C2OC3=C(N1C(=O)NNC(=O)CCSCC4=CC=CO4)C=C(C=C3)Cl
457.929
457.93
10mg
C22H20ClN3O4S
CQBVTZDISUKDSX-UHFFFAOYSA-N
3-chloro-N'-[3-(furan-2-ylmethylsulfanyl)propanoyl]-6H-benzo[b][1,4]benzoxazepine-5-carbohydrazide
9933831
>98%
Videos
Documents