missing translation for 'onlineSavingsMsg'
Learn More

SC 236, Tocris Bioscience™

Selective cyclooxygenase-2 (COX-2) inhibitor

Brand:  Tocris (Bio-Techne) 391910

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 10mg

 View more versions of this product

Product Code. 13433814

  • £173.46 / 10mg

Estimated Shipment date
from Supplier 17-12-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Selective COX-2 inhibitor (IC50 values are 0.005 and 17.8 μM for COX-2 and COX-1 respectively). Displays anti-inflammatory properties and potent antimetastatic activity against both experimental metastases and spontaneous metastases arising following primary tumor excision.
Specifications

Chemical Identifiers

170569-86-5
401.788
4-5-4-chlorophenyl-3-trifluoromethyl-1h-pyrazol-1-yl benzenesulfonamide, 4-5-4-chlorophenyl-3-trifluoromethyl pyrazol-1-yl benzenesulfonamide, d03nzv, cox-2 inhibitors anticancer , pfizer, cox-2 inhibitors anticancer , pharmacia, sc hplc, 4-5 4-chlorophenyl-3-trifluoromethyl-1h-pyrazol-1yl benzenesulfonamide, 4-5-4-chloro-phenyl-3-trifluoromethyl-pyrazol-1-yl-benzenesulfonamide
4-[5-(4-chlorophenyl)-3-(trifluoromethyl)pyrazol-1-yl]benzenesulfonamide
C16H11ClF3N3O2S
NSQNZEUFHPTJME-UHFFFAOYSA-N
9865808
C1=CC(=CC=C1C2=CC(=NN2C3=CC=C(C=C3)S(=O)(=O)N)C(F)(F)F)Cl

Specifications

10mg
170569-86-5
4-5-4-chlorophenyl-3-trifluoromethyl-1h-pyrazol-1-yl benzenesulfonamide, 4-5-4-chlorophenyl-3-trifluoromethyl pyrazol-1-yl benzenesulfonamide, d03nzv, cox-2 inhibitors anticancer , pfizer, cox-2 inhibitors anticancer , pharmacia, sc hplc, 4-5 4-chlorophenyl-3-trifluoromethyl-1h-pyrazol-1yl benzenesulfonamide, 4-5-4-chloro-phenyl-3-trifluoromethyl-pyrazol-1-yl-benzenesulfonamide
C1=CC(=CC=C1C2=CC(=NN2C3=CC=C(C=C3)S(=O)(=O)N)C(F)(F)F)Cl
401.788
401.79
SC 236
C16H11ClF3N3O2S
NSQNZEUFHPTJME-UHFFFAOYSA-N
4-[5-(4-chlorophenyl)-3-(trifluoromethyl)pyrazol-1-yl]benzenesulfonamide
9865808
>99%
Videos
Documents