missing translation for 'onlineSavingsMsg'
Learn More

SC 19220, Tocris Bioscience™

Selective EP1 receptor antagonist

Brand:  Tocris (Bio-Techne) 120650

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13488863

  • £600.29 / 50mg


Estimated Shipment
04-10-2018
Add to basket

Description

Description

Selective EP1 receptor antagonist (IC50 = 6.7 μM for inhibition of [3H]-PGE2 binding to EP1 transfected COS cells).
Specifications

Chemical Identifiers

19395-87-0
331.756
8-Chloro-dibenzo(Z)[b,f][1,4]oxazepine-10(11H)-carboxylic acid, 2-acetylhydrazide
N'-acetyl-3-chloro-6H-benzo[b][1,4]benzoxazepine-5-carbohydrazide
C16H14ClN3O3
KNURFLJTOUGOOQ-UHFFFAOYSA-N
4336830
CC(=O)NNC(=O)N1CC2=CC=CC=C2OC3=C1C=C(C=C3)Cl

Specifications

SC 19220
19395-87-0
C16H14ClN3O3
KNURFLJTOUGOOQ-UHFFFAOYSA-N
N'-acetyl-3-chloro-6H-benzo[b][1,4]benzoxazepine-5-carbohydrazide
4336830
50mg
>99%
8-Chloro-dibenzo(Z)[b,f][1,4]oxazepine-10(11H)-carboxylic acid, 2-acetylhydrazide
CC(=O)NNC(=O)N1CC2=CC=CC=C2OC3=C1C=C(C=C3)Cl
331.756
331.76
Videos