Learn More
SC-10, Tocris Bioscience™
Brand:  Tocris (Bio-Techne) 043010
Code : Z1
Additional Details : Weight : 0.01000kg
Quantity 10mg
 View more versions of this product
Product Code. 13447643
  • £145.53 / 10mg
Estimated Shipment date
from Supplier 19-11-2019
Specifications
Chemical Identifiers
102649-79-6
339.878
5-chloro-n-heptyl-1-naphthalenesulfonamide, tocris-0430, n-m-heptyl-5-chloro-1-naphthalenesulfonamide, 1-naphthalenesulfonamide, 5-chloro-n-heptyl, n-heptyl-5-chloronaphthalene-1-sulfonamide, sc-10, 5-chloro-n-heptylnaphthalene-1-sulfonamide, n-n-heptyl-5-chloro-1-naphthalenesulfonamide
5-chloro-N-heptylnaphthalene-1-sulfonamide
C17H22ClNO2S
MJMJERJFCHYXEM-UHFFFAOYSA-N
5175
CCCCCCCNS(=O)(=O)C1=CC=CC2=C1C=CC=C2Cl
Specifications
10mg
102649-79-6
C17H22ClNO2S
MJMJERJFCHYXEM-UHFFFAOYSA-N
5-chloro-N-heptylnaphthalene-1-sulfonamide
5175
SC-10
>98%
5-chloro-n-heptyl-1-naphthalenesulfonamide, tocris-0430, n-m-heptyl-5-chloro-1-naphthalenesulfonamide, 1-naphthalenesulfonamide, 5-chloro-n-heptyl, n-heptyl-5-chloronaphthalene-1-sulfonamide, sc-10, 5-chloro-n-heptylnaphthalene-1-sulfonamide, n-n-heptyl-5-chloro-1-naphthalenesulfonamide
CCCCCCCNS(=O)(=O)C1=CC=CC2=C1C=CC=C2Cl
339.878
339.88