missing translation for 'onlineSavingsMsg'
Learn More

SBE 13 hydrochloride, Tocris Bioscience™

Potent and selective PLK1 inhibitor

Brand:  RD Systems (Bio-Techne) 4292/50

Code : Z1

Additional Details : Weight : 0.00050kg

Quantity 50mg

 View more versions of this product

Product Code. 13444504

  • £327.81 / 50mg

Estimated Shipment date
from Supplier 18-12-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Selective inhibitor of PLK1 (IC50 values are 200 pM, 875 nM and 66 μM for PLK1, PLK3 and PLK2 respectively). Exhibits no effect on Aurora kinase A activity. Binds and stabilizes the inactive form of PLK1. Delays cell cycle progression, reduces cell proliferation and induces apoptosis in a range of human cancer cell lines. Transiently arrests cells cycle at G0/G1 in primary cells.
Specifications

Chemical Identifiers

1052532-15-6
479.398
sbe13 hydrochloride, sbe 13 hydrochloride, sbe 13 hcl, n-4-6-chloro-3-pyridinyl methoxy-3-methoxyphenyl methyl-3,4-dimethoxybenzeneethanamine hydrochloride, n-4-6-chloropyridin-3-yl methoxy-3-methoxybenzyl-2-3,4-dimethoxyphenyl ethanamine, c24h27cln2o4.clh, sbe-13 hcl;sbe13 hcl, sbe 13 hydrochloride hplc, n-4-6-chloropyridin-3-yl methoxy-3-methoxyphenyl methyl-2-3,4-dimethoxyphenyl ethan-1-amine-hydrogen chloride 1/1
N-[[4-[(6-chloropyridin-3-yl)methoxy]-3-methoxyphenyl]methyl]-2-(3,4-dimethoxyphenyl)ethanamine;hydrochloride
C24H28Cl2N2O4
QBGSVDJLQQXEGG-UHFFFAOYSA-N
11948807
COC1=C(C=C(C=C1)CCNCC2=CC(=C(C=C2)OCC3=CN=C(C=C3)Cl)OC)OC.Cl

Specifications

50mg
1052532-15-6
sbe13 hydrochloride, sbe 13 hydrochloride, sbe 13 hcl, n-4-6-chloro-3-pyridinyl methoxy-3-methoxyphenyl methyl-3,4-dimethoxybenzeneethanamine hydrochloride, n-4-6-chloropyridin-3-yl methoxy-3-methoxybenzyl-2-3,4-dimethoxyphenyl ethanamine, c24h27cln2o4.clh, sbe-13 hcl;sbe13 hcl, sbe 13 hydrochloride hplc, n-4-6-chloropyridin-3-yl methoxy-3-methoxyphenyl methyl-2-3,4-dimethoxyphenyl ethan-1-amine-hydrogen chloride 1/1
COC1=C(C=C(C=C1)CCNCC2=CC(=C(C=C2)OCC3=CN=C(C=C3)Cl)OC)OC.Cl
479.398
479.4
SBE 13 hydrochloride
C24H28Cl2N2O4
QBGSVDJLQQXEGG-UHFFFAOYSA-N
N-[[4-[(6-chloropyridin-3-yl)methoxy]-3-methoxyphenyl]methyl]-2-(3,4-dimethoxyphenyl)ethanamine;hydrochloride
11948807
>99%
Videos
Documents