Learn More
Tocris Bioscience™ SB 525334
Selective inhibitor of TGF-βRI
Brand:  Tocris Bioscience™ 321150
Code : Z1
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 13432574
  • £842.31 / 50mg
Estimated Shipment date
from Supplier 02-04-2020
Description
Description
Selective inhibitor of transforming growth factor-β receptor I (ALK5, TGF-βRI) (IC50 = 14.3 nM). Inhibits TGF-β1-induced smad2/3 nuclear localization and TGF-βRI-induced mRNA expression in kidney cells. Attenuates bleomycin-induced pulmonary fibrosis.
Specifications
Specifications
SB 525334
>97
50mg
SB 525334
343.42