missing translation for 'onlineSavingsMsg'
Learn More

SB 452533, Tocris Bioscience™

Potent TRPV1 receptor antagonist

Brand:  Tocris (Bio-Techne) 326550

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13462684

  • £431.62 / 50mg


Estimated Shipment
04-03-2019
Add to basket

Description

Description

Potent TRPV1 antagonist against capsaicin (pKb = 7.7), noxious heat and acid-mediated (pIC50 = 7.0) receptor activation (pKi = 6.22 at the recombinant hTRPV1 receptor). Exhibits analgesic properties.
Specifications

Chemical Identifiers

459429-39-1
376.298
n-2-bromophenyl-n'-2-ethyl 3-methylphenyl amino ethyl-urea, n-2-bromophenyl-n'-2-ethyl 3-methylphenyl amino ethyl-urea, d0u3wk, 1-2-bromophenyl-3-2-ethyl 3-methylphenyl amino ethyl urea, 3-2-bromophenyl-1-2-ethyl 3-methylphenyl amino ethyl urea, n-2-bromophenyl-n'-2-n-ethyl-n-3-methylphenyl amino ethyl urea
1-(2-bromophenyl)-3-[2-(N-ethyl-3-methylanilino)ethyl]urea
C18H22BrN3O
IFJYEGJUQIBBQV-UHFFFAOYSA-N
9842609
CCN(CCNC(=O)NC1=CC=CC=C1Br)C2=CC=CC(=C2)C

Specifications

SB 452533
459429-39-1
n-2-bromophenyl-n'-2-ethyl 3-methylphenyl amino ethyl-urea, n-2-bromophenyl-n'-2-ethyl 3-methylphenyl amino ethyl-urea, d0u3wk, 1-2-bromophenyl-3-2-ethyl 3-methylphenyl amino ethyl urea, 3-2-bromophenyl-1-2-ethyl 3-methylphenyl amino ethyl urea, n-2-bromophenyl-n'-2-n-ethyl-n-3-methylphenyl amino ethyl urea
CCN(CCNC(=O)NC1=CC=CC=C1Br)C2=CC=CC(=C2)C
376.298
376.29
50mg
C18H22BrN3O
IFJYEGJUQIBBQV-UHFFFAOYSA-N
1-(2-bromophenyl)-3-[2-(N-ethyl-3-methylanilino)ethyl]urea
9842609
>98%
Videos