missing translation for 'onlineSavingsMsg'
Learn More

SB 399885 hydrochloride, Tocris Bioscience™

Potent and selective 5-HT6 antagonist

Brand:  Tocris (Bio-Techne) 318950

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13442394

  • £815.85 / 50mg

Estimated Shipment date
from Supplier 25-10-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Potent, brain penetrant, orally active 5-HT6 antagonist. Displays > 200-fold selectivity for 5-HT6 over other 5-HT receptors (pKi values are 9.11, 8.81 and 9.02 for human recombinant, native rat and native human 5-HT6 receptors respectively).
Specifications

Chemical Identifiers

402713-81-9
482.801
sb 399885 hydrochloride, sb-399885 hydrochloride, n-3,5-dichloro-2-methoxyphenyl-4-methoxy-3-1-piperazinyl benzenesulfonamide hydrochloride, n-3,5-dichloro-2-methoxy-phenyl-4-methoxy-3-piperazin-1-yl-benzenesulfonamide hydrochloride, n-3,5-dichloro-2-methoxyphenyl-4-methoxy-3-piperazin-1-yl benzenesulfonamide hydrochloride
N-(3,5-dichloro-2-methoxyphenyl)-4-methoxy-3-piperazin-1-ylbenzenesulfonamide;hydrochloride
C18H22Cl3N3O4S
RNKCEBCFUSXSQE-UHFFFAOYSA-N
10277652
COC1=C(C=C(C=C1)S(=O)(=O)NC2=CC(=CC(=C2OC)Cl)Cl)N3CCNCC3.Cl

Specifications

50mg
402713-81-9
sb 399885 hydrochloride, sb-399885 hydrochloride, n-3,5-dichloro-2-methoxyphenyl-4-methoxy-3-1-piperazinyl benzenesulfonamide hydrochloride, n-3,5-dichloro-2-methoxy-phenyl-4-methoxy-3-piperazin-1-yl-benzenesulfonamide hydrochloride, n-3,5-dichloro-2-methoxyphenyl-4-methoxy-3-piperazin-1-yl benzenesulfonamide hydrochloride
COC1=C(C=C(C=C1)S(=O)(=O)NC2=CC(=CC(=C2OC)Cl)Cl)N3CCNCC3.Cl
482.801
482.81
SB 399885 hydrochloride
C18H22Cl3N3O4S
RNKCEBCFUSXSQE-UHFFFAOYSA-N
N-(3,5-dichloro-2-methoxyphenyl)-4-methoxy-3-piperazin-1-ylbenzenesulfonamide;hydrochloride
10277652
>99%
Videos
Documents