Learn More
SB 366791, Tocris Bioscience™
Potent, selective, competitive TRPV1 antagonist
Brand:  Tocris (Bio-Techne) 1615/10
Code : Z1
Additional Details : Weight : 0.01000kg
Quantity 10mg
 View more versions of this product
Product Code. 13449803
  • £160.23 / 10mg
Call for latest availability
+44 (0)1509 555 500
Description
Description
Potent, selective and competitive vanilloid TRPV1 receptor antagonist (pA2 = 7.71 at hVR1); antagonizes hTRPV1 receptors activated by agonists, noxious heat, but not protons. Displays selectivity over a wide range of receptors and systems including CB1 and CB2 receptors, voltage-gated Ca2+ channels and the hyperpolarization-activated current (Ih). Also available as part of the Vanilloid TRPV1 Receptor Tocriset™ .
Specifications
Chemical Identifiers
472981-92-3
287.743
2e-3-4-chlorophenyl-n-3-methoxyphenyl prop-2-enamide, n-3-methoxyphenyl-4-chlorocinnamide, n-3-methoxyphenyl-4-chlorocinnamanilide, e-3-4-chlorophenyl-n-3-methoxyphenyl acrylamide, ftalidap 27, 3-4-chlorophenyl-n-3-methoxyphenyl prop-2-enamide, 2-propenamide, 3-4-chlorophenyl-n-3-methoxyphenyl, sb hplc , powder, e-3-4-chlorophenyl-n-3-methoxyphenyl prop-2-enamide
(E)-3-(4-chlorophenyl)-N-(3-methoxyphenyl)prop-2-enamide
C16H14ClNO2
RYAMDQKWNKKFHD-JXMROGBWSA-N
667594
COC1=CC=CC(=C1)NC(=O)C=CC2=CC=C(C=C2)Cl
Specifications
10mg
472981-92-3
2e-3-4-chlorophenyl-n-3-methoxyphenyl prop-2-enamide, n-3-methoxyphenyl-4-chlorocinnamide, n-3-methoxyphenyl-4-chlorocinnamanilide, e-3-4-chlorophenyl-n-3-methoxyphenyl acrylamide, ftalidap 27, 3-4-chlorophenyl-n-3-methoxyphenyl prop-2-enamide, 2-propenamide, 3-4-chlorophenyl-n-3-methoxyphenyl, sb hplc , powder, e-3-4-chlorophenyl-n-3-methoxyphenyl prop-2-enamide
COC1=CC=CC(=C1)NC(=O)C=CC2=CC=C(C=C2)Cl
287.743
287.75
SB 366791
C16H14ClNO2
RYAMDQKWNKKFHD-JXMROGBWSA-N
(E)-3-(4-chlorophenyl)-N-(3-methoxyphenyl)prop-2-enamide
667594
>99%