Learn More
Tocris Bioscience™ SB 334867
Selective non-peptide OX1 antagonist
Brand:  Tocris Bioscience™ 1960/50
Code : Z1
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 13460304
  • £645.09 / 50mg
Estimated Shipment
28-02-2019
Add to basket
Description
Description
Selective non-peptide orexin OX1 receptor antagonist. pKb values are 7.2 and < 5 for inhibition of intracellular Ca2+ release in CHO cells expressing human OX1 and OX2 receptors respectively. Blocks orexin-A induced grooming and feeding following systemic administration in vivo.
Specifications
Specifications
N-(2-Methyl-6-benzoxazolyl)-N'-1,5-naphthyridin-4-yl urea
>99%
Observed MW: 319.32
Soluble to 100mM with gentle warming in DMSO
792173-99-0
C17H13N5O2