missing translation for 'onlineSavingsMsg'
Learn More

SB 269970 hydrochloride, Tocris Bioscience™

Potent and selective 5-HT7 antagonist; brain penetrant

Brand:  Tocris (Bio-Techne) 1612/10

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 10mg

 View more versions of this product

Product Code. 13469793

  • £208.74 / 10mg

Estimated Shipment date
from Supplier 06-04-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Potent and selective 5-HT7 receptor antagonist (pKi values are 8.9, 7.2 and 6.0 for 5-HT7A, 5-HT5A and 5-HT1B and < 6.0 for 5-HT1A, 5-HT1D, 5-HT1E, 5-HT1F, 5-HT2A, 5-HT2B, 5-HT2C, 5-HT4 and 5-HT6 receptors respectively). Brain penetrant in vivo.
Specifications

Chemical Identifiers

261901-57-9
388.951
sb-269970 hydrochloride, sb 269970 hydrochloride, r-3-2-2-4-methylpiperidin-1-yl ethyl pyrrolidin-1-yl sulfonyl phenol hydrochloride, r-3-2-2-4-methylpiperidin-1-yl ethyl pyrrolidine-1-sulfonyl phenol hydrochloride, 3-2r-2-2-4-methylpiperidin-1-yl ethyl pyrrolidin-1-ylsulfonyl phenol hydrochloride, d0bg5h, sb-269970 hydrochloride, powder hplc, r-3-2-2-4-methylpiperidin-1-yl ethyl pyrrolidin-1-yl sulfonyl phenol hcl, r-3-2-2-4-methyl-piperidin-1-yl ethyl pyrrolidine-1-sulfonyl phenol hydrochloride
3-[(2R)-2-[2-(4-methylpiperidin-1-yl)ethyl]pyrrolidin-1-yl]sulfonylphenol;hydrochloride
C18H29ClN2O3S
XQCJOYZLWFNDIO-PKLMIRHRSA-N
11957684
CC1CCN(CC1)CCC2CCCN2S(=O)(=O)C3=CC=CC(=C3)O.Cl

Specifications

10mg
261901-57-9
sb-269970 hydrochloride, sb 269970 hydrochloride, r-3-2-2-4-methylpiperidin-1-yl ethyl pyrrolidin-1-yl sulfonyl phenol hydrochloride, r-3-2-2-4-methylpiperidin-1-yl ethyl pyrrolidine-1-sulfonyl phenol hydrochloride, 3-2r-2-2-4-methylpiperidin-1-yl ethyl pyrrolidin-1-ylsulfonyl phenol hydrochloride, d0bg5h, sb-269970 hydrochloride, powder hplc, r-3-2-2-4-methylpiperidin-1-yl ethyl pyrrolidin-1-yl sulfonyl phenol hcl, r-3-2-2-4-methyl-piperidin-1-yl ethyl pyrrolidine-1-sulfonyl phenol hydrochloride
CC1CCN(CC1)CCC2CCCN2S(=O)(=O)C3=CC=CC(=C3)O.Cl
388.951
388.95
SB 269970 hydrochloride
C18H29ClN2O3S
XQCJOYZLWFNDIO-PKLMIRHRSA-N
3-[(2R)-2-[2-(4-methylpiperidin-1-yl)ethyl]pyrrolidin-1-yl]sulfonylphenol;hydrochloride
11957684
>98%
Videos
Documents