missing translation for 'onlineSavingsMsg'
Learn More

SB 265610, Tocris Bioscience™

Potent CXCR2 antagonist

Brand:  Tocris (Bio-Techne) 272410

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 10mg

 View more versions of this product

Product Code. 13451804

  • £219.03 / 10mg

Estimated Shipment date
from Supplier 30-03-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Potent CXCR2 antagonist that inhibits CINC-1-mediated but not C5a-mediated Ca2+ mobilization (IC50 values are 3.4 and 6800 nM respectively). Inhibits CINC-induced chemotaxis and attenuates neutrophil accumulation in inflammatory lung injury in vivo.
Specifications

Chemical Identifiers

211096-49-0
357.171
chembl38182, n-2-bromophenyl-n'-7-cyano-1h-benzotriazol-4-yl urea, 1-2-bromophenyl-3-4-cyano-1h-benzo d 1,2,3 triazol-7-yl urea, 1-2-bromophenyl-3-7-cyano-1h-1,2,3-benzotriazol-4-yl urea, d02jxg, sb hplc, 1-2-bromo-phenyl-3-7-cyano-3h-benzotriazol-4-yl-urea, urea, n-2-bromophenyl-n'-4-cyano-1h-benzotriazol-7-yl, 1-2-bromophenyl-3-7-cyano-2h-1,2,3-benzotriazol-4-yl urea, 1-2-bromophenyl-3-7-cyano-3h-1,2,3-benzotriazol-4-yl urea
1-(2-bromophenyl)-3-(7-cyano-2H-benzotriazol-4-yl)urea
C14H9BrN6O
SEDUMQWZEOMXSO-UHFFFAOYSA-N
9841667
C1=CC=C(C(=C1)NC(=O)NC2=CC=C(C3=NNN=C23)C#N)Br

Specifications

10mg
211096-49-0
chembl38182, n-2-bromophenyl-n'-7-cyano-1h-benzotriazol-4-yl urea, 1-2-bromophenyl-3-4-cyano-1h-benzo d 1,2,3 triazol-7-yl urea, 1-2-bromophenyl-3-7-cyano-1h-1,2,3-benzotriazol-4-yl urea, d02jxg, sb hplc, 1-2-bromo-phenyl-3-7-cyano-3h-benzotriazol-4-yl-urea, urea, n-2-bromophenyl-n'-4-cyano-1h-benzotriazol-7-yl, 1-2-bromophenyl-3-7-cyano-2h-1,2,3-benzotriazol-4-yl urea, 1-2-bromophenyl-3-7-cyano-3h-1,2,3-benzotriazol-4-yl urea
C1=CC=C(C(=C1)NC(=O)NC2=CC=C(C3=NNN=C23)C#N)Br
357.171
357.16
SB 265610
C14H9BrN6O
SEDUMQWZEOMXSO-UHFFFAOYSA-N
1-(2-bromophenyl)-3-(7-cyano-2H-benzotriazol-4-yl)urea
9841667
>98%
Videos
Documents