Learn More
Tocris Bioscience™ SB 224289 hydrochloride
Brand:  Tocris Bioscience™ 122110
Code : Z1
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 13488913
  • £173.00 / 10mg
Estimated Shipment
05-07-2018
Add to basket
Description
Description
Selective 5-HT1B receptor antagonist (pKi = 8.2). Displays > 60-fold selectivity over 5-HT1D, 5-HT1A, 5-HT1E, 5-HT1F, 5-HT2A and 5-HT2C receptors in radioligand binding and functional assays. Centrally active following oral administration in vivo.
Specifications
Specifications
1'-Methyl-5-[[2'-methyl-4'-(5-methyl-1,2,4-oxadiazol-3-yl)biphenyl-4-yl]carbonyl]-2,3,6,7-tetrahydrospiro[furo[2,3-f]indole-3,4'-piperidine hydrochloride
>95%
Observed MW: 557.09
Soluble to 10mM with gentle warming in DMSO
180084-26-8
C32H32N4O3.HCl