missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SB 222200, Tocris Bioscience™
Potent, selective non-peptide NK3 antagonist. Brain penetrant
Brand: Tocris (Bio-Techne) 1393/10
Code : Z1
Additional Details : Weight : 0.01000kg
Quantity | 10mg |
---|
Description
Potent and selective non-peptide NK3 receptor antagonist (Ki values are 4.4, > 100,000 and 250 nM for human NK3, NK1 and NK2 receptors respectively). Antihypertensive in vivo. Brain penetrant.Chemical Identifiers
174635-69-9 | |
380.491 | |
3-methyl-2-phenyl-n-1s-1-phenylpropyl quinoline-4-carboxamide, chembl10284, s-3-methyl-2-phenyl-n-1-phenylpropyl-4-quinolinecarboxamide, 3-methyl-2-phenyl-n-1s-1-phenylpropyl-4-quinolinecarboxamide, s-3-methyl-2-phenyl-n-1-phenylpropyl quinoline-4-carboxamide, 4-quinolinecarboxamide, 3-methyl-2-phenyl-n-1s-1-phenylpropyl, 4-quinolinecarboxamide, 3-methyl-2-phenyl-n-1s-1-phenylpropyl, tocris-1393, lopac-s-5192, d00vpt | |
3-methyl-2-phenyl-N-[(1S)-1-phenylpropyl]quinoline-4-carboxamide |
C26H24N2O | |
MQNYRKWJSMQECI-QFIPXVFZSA-N | |
6604009 | |
CCC(C1=CC=CC=C1)NC(=O)C2=C(C(=NC3=CC=CC=C32)C4=CC=CC=C4)C |
Specifications
SB 222200 | |
174635-69-9 | |
3-methyl-2-phenyl-n-1s-1-phenylpropyl quinoline-4-carboxamide, chembl10284, s-3-methyl-2-phenyl-n-1-phenylpropyl-4-quinolinecarboxamide, 3-methyl-2-phenyl-n-1s-1-phenylpropyl-4-quinolinecarboxamide, s-3-methyl-2-phenyl-n-1-phenylpropyl quinoline-4-carboxamide, 4-quinolinecarboxamide, 3-methyl-2-phenyl-n-1s-1-phenylpropyl, 4-quinolinecarboxamide, 3-methyl-2-phenyl-n-1s-1-phenylpropyl, tocris-1393, lopac-s-5192, d00vpt | |
CCC(C1=CC=CC=C1)NC(=O)C2=C(C(=NC3=CC=CC=C32)C4=CC=CC=C4)C | |
380.491 | |
380.48 |
10mg | |
C26H24N2O | |
MQNYRKWJSMQECI-QFIPXVFZSA-N | |
3-methyl-2-phenyl-N-[(1S)-1-phenylpropyl]quinoline-4-carboxamide | |
6604009 | |
>98% |