missing translation for 'onlineSavingsMsg'
Learn More

SB 221284, Tocris Bioscience™

Potent, selective 5-HT2C/2B antagonist

Brand:  Tocris (Bio-Techne) 137910

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 10mg

 View more versions of this product

Product Code. 13439263

  • £169.05 / 10mg

Estimated Shipment date
from Supplier 19-11-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Potent, selective 5-HT2C/2B receptor antagonist. pKi values are 6.4, 7.9 and 8.6 for 5-HT2A, 2B and 2C receptors respectively. Centrally active upon systemic administration in vivo.
Specifications

Chemical Identifiers

196965-14-7
353.363
tocris-1379, 2,3-dihydro-5-methylthio-n-3-pyridinyl-6-trifluoromethyl-1h-indole-1-carboxamide, 5-methylsulfanyl-n-pyridin-3-yl-6-trifluoromethyl-2,3-dihydroindole-1-carboxamide, d0dy5g, gtpl191, 5-methylthio-1-3-pyridylcarbamoyl-6-trifluoromethyl indoline, 5-methylthio-n-pyridin-3-yl-6-trifluoromethyl indoline-1-carboxamide, 5-methylsulfanyl-n-3-pyridyl-6-trifluoromethyl indoline-1-carboxamide
CHEBI:8978
CSC1=C(C=C2C(=C1)CCN2C(=O)NC3=CN=CC=C3)C(F)(F)F
C16H14F3N3OS
OQZOXHCRSXYSPM-UHFFFAOYSA-N
443389
5-methylsulfanyl-N-pyridin-3-yl-6-(trifluoromethyl)-2,3-dihydroindole-1-carboxamide

Specifications

10mg
196965-14-7
tocris-1379, 2,3-dihydro-5-methylthio-n-3-pyridinyl-6-trifluoromethyl-1h-indole-1-carboxamide, 5-methylsulfanyl-n-pyridin-3-yl-6-trifluoromethyl-2,3-dihydroindole-1-carboxamide, d0dy5g, gtpl191, 5-methylthio-1-3-pyridylcarbamoyl-6-trifluoromethyl indoline, 5-methylthio-n-pyridin-3-yl-6-trifluoromethyl indoline-1-carboxamide, 5-methylsulfanyl-n-3-pyridyl-6-trifluoromethyl indoline-1-carboxamide
CSC1=C(C=C2C(=C1)CCN2C(=O)NC3=CN=CC=C3)C(F)(F)F
353.363
CHEBI:8978
>99%
SB 221284
C16H14F3N3OS
OQZOXHCRSXYSPM-UHFFFAOYSA-N
5-methylsulfanyl-N-pyridin-3-yl-6-(trifluoromethyl)-2,3-dihydroindole-1-carboxamide
443389
353.36
Videos
Documents