missing translation for 'onlineSavingsMsg'
Learn More

SB 218078, Tocris Bioscience™

Chk1 inhibitor

Brand:  RD Systems (Bio-Techne) 25601

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 1mg

 View more versions of this product

Product Code. 13441474

  • £144.06 / 1mg

Estimated Shipment date
from Supplier 18-12-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Inhibitor of checkpoint kinase 1 (Chk1) that displays selectivity over other protein kinases (IC50 values are 15, 250 and 1000 nM for Chk1, cdc2 and PKC respectively). Abrogates G2 cell cycle arrest caused by γ-irradiation and topoisomerase I inhibition. Potentiates cytotoxicity of DNA-damaging drugs, enhancing the efficacy of some chemotherapeutics.
Specifications

Chemical Identifiers

135897-06-2
393.402
9,10,11,12,-tetrahydro-9,12-epoxy-1h-diindolo 1,2,3-fg:3′,2′,1′-kl pyrrolo 3,4-i 1,6 benzodiazocine-1,3 2h-dione, d00kdf, 5,6,7,8-tetrahydro-13h-16-oxa-4b,8a,14-triaza-5,8-methanodibenzo b,h cycloocta jkl cyclopenta e-as-indacene-13,15 14h-dione, 9,10,11,12-tetrahydro-9,12-epoxy-1h-diindolo 1,2,3-fg:3',2',1'-kl pyrrolo 3,4-i 1,6 benzodiazocine-1,3 2h-dione, 9,12-epoxy-1h-diindolo 1,2,3-fg:3',2',1'-kl pyrrolo 3,4-i 1,6 benzodiazocine-1,3 2h-dione, 9,10,11, 9,12-epoxy-1h-diindolo 1,2,3-fg:3',2',1'-kl pyrrolo 3,4-i 1,6 benzodiazocine-1,3 2h-dione,9,10,11,12-tetrahydro
C1CC2N3C4=CC=CC=C4C5=C6C(=C7C8=CC=CC=C8N(C1O2)C7=C53)C(=O)NC6=O
C24H15N3O3
OTPNDVKVEAIXTI-UHFFFAOYSA-N
3387354

Specifications

1mg
135897-06-2
9,10,11,12,-tetrahydro-9,12-epoxy-1h-diindolo 1,2,3-fg:3′,2′,1′-kl pyrrolo 3,4-i 1,6 benzodiazocine-1,3 2h-dione, d00kdf, 5,6,7,8-tetrahydro-13h-16-oxa-4b,8a,14-triaza-5,8-methanodibenzo b,h cycloocta jkl cyclopenta e-as-indacene-13,15 14h-dione, 9,10,11,12-tetrahydro-9,12-epoxy-1h-diindolo 1,2,3-fg:3',2',1'-kl pyrrolo 3,4-i 1,6 benzodiazocine-1,3 2h-dione, 9,12-epoxy-1h-diindolo 1,2,3-fg:3',2',1'-kl pyrrolo 3,4-i 1,6 benzodiazocine-1,3 2h-dione, 9,10,11, 9,12-epoxy-1h-diindolo 1,2,3-fg:3',2',1'-kl pyrrolo 3,4-i 1,6 benzodiazocine-1,3 2h-dione,9,10,11,12-tetrahydro
C1CC2N3C4=CC=CC=C4C5=C6C(=C7C8=CC=CC=C8N(C1O2)C7=C53)C(=O)NC6=O
3387354
>98%
SB 218078
C24H15N3O3
OTPNDVKVEAIXTI-UHFFFAOYSA-N
393.402
393.39
Videos
Documents