missing translation for 'onlineSavingsMsg'
Learn More

SB 216641 hydrochloride, Tocris Bioscience™

Selective h5-HT1B antagonist

Brand:  Tocris (Bio-Techne) 124210

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 10mg

 View more versions of this product

Product Code. 13458953

  • £191.40 / 10mg


Estimated Shipment
29-10-2018
Add to basket

Description

Description

A selective h5-HT1B antagonist with approximately 25-fold selectivity over h5-HT1D and little or no affinity for a range of other receptor types.
Specifications

Chemical Identifiers

193611-67-5
523.03
sb 216641 hydrochloride, n-2-dimethylamino ethyl-n-4-2-phenylethyl amino methyl 1,1-biphenyl-4-yl methyl cyclopenta, n-3-3-dimethylamino ethoxy-4-methoxyphenyl-2'-methyl-4'-5-methyl-1,2,4-oxadiazol-3-yl-1,1'-biphenyl-4-carboxamide hydrochloride, n-3-2-dimethylamino ethoxy-4-methoxyphenyl-2'-methyl-4'-5-methyl-1,2,4-oxadiazol-3-yl biphenyl-4-carboxamide hydrochloride
N-[3-[2-(dimethylamino)ethoxy]-4-methoxyphenyl]-4-[2-methyl-4-(5-methyl-1,2,4-oxadiazol-3-yl)phenyl]benzamide;hydrochloride
C28H31ClN4O4
JPBMDMNORXKGHZ-UHFFFAOYSA-N
10392025
CC1=C(C=CC(=C1)C2=NOC(=N2)C)C3=CC=C(C=C3)C(=O)NC4=CC(=C(C=C4)OC)OCCN(C)C.Cl

Specifications

SB 216641 hydrochloride
193611-67-5
sb 216641 hydrochloride, n-2-dimethylamino ethyl-n-4-2-phenylethyl amino methyl 1,1-biphenyl-4-yl methyl cyclopenta, n-3-3-dimethylamino ethoxy-4-methoxyphenyl-2'-methyl-4'-5-methyl-1,2,4-oxadiazol-3-yl-1,1'-biphenyl-4-carboxamide hydrochloride, n-3-2-dimethylamino ethoxy-4-methoxyphenyl-2'-methyl-4'-5-methyl-1,2,4-oxadiazol-3-yl biphenyl-4-carboxamide hydrochloride
CC1=C(C=CC(=C1)C2=NOC(=N2)C)C3=CC=C(C=C3)C(=O)NC4=CC(=C(C=C4)OC)OCCN(C)C.Cl
523.03
523.03
10mg
C28H31ClN4O4
JPBMDMNORXKGHZ-UHFFFAOYSA-N
N-[3-[2-(dimethylamino)ethoxy]-4-methoxyphenyl]-4-[2-methyl-4-(5-methyl-1,2,4-oxadiazol-3-yl)phenyl]benzamide;hydrochloride
10392025
>98%
Videos