missing translation for 'onlineSavingsMsg'
Learn More

SB 203580, Tocris Bioscience™

Selective inhibitor of p38 MAPK

Brand:  Tocris (Bio-Techne) 1202/10

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 10mg

 View more versions of this product

Product Code. 13488853

  • £209.88 / 10mg


Estimated Shipment
27-09-2018
Add to basket

Description

Description

Selective inhibitor of p38 MAPK (IC50 values are 50 and 500 nM for SAPK2a/p38 and SAPK2b/p38β2 respectively). Displays 100-500-fold selectivity over LCK, GSK-3β and PKBα. Shown to inhibit IL-2-induced T cell proliferation, cyclooxygenase-1 and -2, and thromboxane synthase. Enhances clonal growth of skin epithelial progenitor cells; stimulates neural stem cell (NSC) proliferation. Essential component of medium for maintaining stem cells in naive pluripotent state. Part of the MAPK Cascade Inhibitor Tocriset™ and MAPK Inhibitor Tocriset™ . Water-soluble salt SB 203580 hydrochloride (Cat. No. 1402) also available.
Specifications

Chemical Identifiers

152121-47-6
377.437
4-[5-(4-Fluorophenyl)-2-[4-(methylsulfonyl)phenyl]-1H-imidazol-4-yl]pyridine
C21H16FN3OS
CDMGBJANTYXAIV-UHFFFAOYSA-N
176155

Specifications

SB 203580
152121-47-6
C21H16FN3OS
CDMGBJANTYXAIV-UHFFFAOYSA-N
4-[4-(4-fluorophenyl)-2-(4-methylsulfinylphenyl)-1H-imidazol-5-yl]pyridine
176155
10mg
>98%
4-[5-(4-Fluorophenyl)-2-[4-(methylsulfonyl)phenyl]-1H-imidazol-4-yl]pyridine
CS(=O)C1=CC=C(C=C1)C2=NC(=C(N2)C3=CC=NC=C3)C4=CC=C(C=C4)F
377.437
377.44
Videos