Learn More
SB 203186 hydrochloride, Tocris Bioscience™
5-HT4 antagonist
Brand:  Tocris (Bio-Techne) 078550
Code : Z1
Additional Details : Weight : 0.01000kg
Quantity 50mg
 View more versions of this product
Product Code. 13458003
  • £349.81 / 50mg
Estimated Shipment
06-03-2019
Add to basket
Description
Description
Potent 5-HT4 receptor antagonist with high affinity for human atrial 5-HT4 receptors.
Specifications
Chemical Identifiers
207572-69-8
C16H20N2O2·HCl
1-Piperidinylethyl-1H-indole-3-carboxylate hydrochloride
Specifications
SB 203186 hydrochloride
207572-69-8
1-Piperidinylethyl-1H-indole-3-carboxylate hydrochloride
>99%
50mg
C16H20N2O2·HCl
308.81
Videos