missing translation for 'onlineSavingsMsg'
Learn More

SB 200646 hydrochloride, Tocris Bioscience™

5-HT2C/2B antagonist

Brand:  Tocris (Bio-Techne) 137110

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 10mg

 View more versions of this product

Product Code. 13469243

  • £130.83 / 10mg

Estimated Shipment date
from Supplier 27-03-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

5-HT2C/2B receptor antagonist, selective over 5-HT1A. Affinities are 7.4 (pA2), 6.9 (pKi) and 5.2 (pKi) for 5-HT2B, 2C and 2A respectively. Orally active in vivo.
Specifications

Chemical Identifiers

143797-62-0
302.762
sb 200646 hydrochloride, sb-200646 hydrochloride, 3-1-methylindol-5-yl-1-pyridin-3-yl urea hydrochloride, 1-1-methylindol-5-yl-3-pyridin-3-ylurea hydrochloride
1-(1-methylindol-5-yl)-3-pyridin-3-ylurea;hydrochloride
C15H15ClN4O
IGRYPUQJEDJLHC-UHFFFAOYSA-N
5311422
CN1C=CC2=C1C=CC(=C2)NC(=O)NC3=CN=CC=C3.Cl

Specifications

10mg
143797-62-0
sb 200646 hydrochloride, sb-200646 hydrochloride, 3-1-methylindol-5-yl-1-pyridin-3-yl urea hydrochloride, 1-1-methylindol-5-yl-3-pyridin-3-ylurea hydrochloride
CN1C=CC2=C1C=CC(=C2)NC(=O)NC3=CN=CC=C3.Cl
302.762
302.76
SB 200646 hydrochloride
C15H15ClN4O
IGRYPUQJEDJLHC-UHFFFAOYSA-N
1-(1-methylindol-5-yl)-3-pyridin-3-ylurea;hydrochloride
5311422
>98%
Videos
Documents