Learn More
SASH1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA556304
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: LLSAKSSTEP SLKSFSRNQL GNYPTLPLMK SGDALKQGQE EGRLGGGLAP DTSKSCDPPG VTGLNKNRRS LPVSICRSCE TLEGPQTVDT WPRSH Highest antigen sequence identity to the following orthologs: Mouse - 78%, Rat - 76%.
SASH1 Polyclonal antibody specifically detects SASH1 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin).Specifications
SASH1 | |
Polyclonal | |
Unconjugated | |
SASH1 | |
1100001C18Rik; 2500002E12Rik; A330076K04Rik; dJ323M4; dJ323M4.1; KIAA0790; mKIAA0790; PEPE1; Proline-glutamate repeat-containing protein; putative adapter and scaffold protein 1; RGD1566017; SAM and SH3 domain containing 1; SAM and SH3 domain-containing protein 1; Sash1; SH3D6A | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
23328 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin) | |
0.5 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O94885 | |
SASH1 | |
Recombinant protein corresponding to Human SASH1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only